MilliporeSigma
  • Stereoselective synthesis of trisubstituted alkenylboranes by palladium-catalysed reaction of alkynyltriarylborates with aryl halides.
Stereoselective synthesis of trisubstituted alkenylboranes by palladium-catalysed reaction of alkynyltriarylborates with aryl halides.
Chemical communications (Cambridge, England) (2007-10-25)
Naoki Ishida, Tomoya Miura, Masahiro Murakami
ABSTRACT
The palladium-catalysed reaction of alkynyltriarylborates with aryl halides afforded trisubstituted alkenylboranes, in which two different aryl groups were installed across the carbon-carbon double bond in a cis arrangement.
Sigma-Aldrich
3-Bromotoluene, 98%
Pricing and availability is not currently available.