MilliporeSigma
  • Complete deletion of the AZFb interval from the Y chromosome in an oligozoospermic man.
Complete deletion of the AZFb interval from the Y chromosome in an oligozoospermic man.
Human reproduction (Oxford, England) (2010-08-19)
Guy Longepied, Noemie Saut, Isabelle Aknin-Seifer, Rachel Levy, Anne-Marie Frances, Catherine Metzler-Guillemain, Marie-Roberte Guichaoua, Michael J Mitchell
ABSTRACT
Deletion of the entire AZFb interval from the Y chromosome is strictly associated with azoospermia arising from maturation arrest during meiosis. Here, we describe the exceptional case of an oligozoospermic man, 13-1217, with an AZFb + c (P5/distal-P1) deletion. Through the characterization of this patient, and two AZFb (P5/proximal-P1) patients with maturation arrest, we have explored three possible explanations for his exceptionally progressive spermatogenesis. We have determined the precise breakpoints of the deletion in 13-1217, and shown that 13-1217 is deleted for more AZFb material than one of the AZFb-deleted men (13-5349). Immunocytochemical analysis of spermatocytes with an antibody against a synaptonemal complex component indicates synapsis to be largely unaffected in 13-1217, in contrast to 13-5349 where extended asynapsis is frequent. Using PCR-based analyses of RNA and DNA from the same testicular biopsy, we show that 13-1217 expresses post-meiotic germ cell markers in the absence of genomic DNA and transcripts from the AZFb and AZFc intervals. We have determined the Y chromosome haplogroup of 13-1217 to be HgL-M185. Our results indicate that the post-meiotic spermatogenesis in 13-1217 is not a consequence of mosaicism or retention of a key AZFb gene. On the contrary, since the Hg-L Y chromosome carried by 13-1217 is uncommon in Western Europe, a Y-linked modifier locus remains a possible explanation for the oligozoospermia observed in patient 13-1217. Further cases must now be studied to understand how germ cells complete spermatogenesis in the absence of the AZFb interval.