MilliporeSigma
  • Di-2-pyridyl ketone oxime in copper chemistry: di-, tri-, penta- and hexanuclear complexes.
Di-2-pyridyl ketone oxime in copper chemistry: di-, tri-, penta- and hexanuclear complexes.
Dalton transactions (Cambridge, England : 2003) (2007-06-20)
Tereza Afrati, Curtis M Zaleski, Catherine Dendrinou-Samara, Gellert Mezei, Jeff W Kampf, Vincent L Pecoraro, Dimitris P Kessissoglou
ABSTRACT
The use of di-2-pyridyl ketone oxime (Hpko)/X- "blends" (X- = OH-, Cl-, ClO4-) in copper chemistry has yielded neutral binuclear and cationic trinuclear, pentanuclear or hexanuclear complexes. Various synthetic procedures have led to the synthesis of compounds [Cu5(pko)7].[ClO4]3.2CH3OH.2H2O (1), [Cu3(pko)3(OH)(Cl)]2[Ph4B]2.4DMF.2H2O (2), [Cu2(pko)4] (3), {[Cu6(pko)6ClO4(CH3CN)6][Cu6(pko)6(ClO4)3(CH3CN)4]}.8ClO4.14CH3CN.H2O (4). The structures of the complexes have been determined by single-crystal X-ray crystallography.