MilliporeSigma
  • Targeted disruption of the GATA3 gene causes severe abnormalities in the nervous system and in fetal liver haematopoiesis.
Targeted disruption of the GATA3 gene causes severe abnormalities in the nervous system and in fetal liver haematopoiesis.
Nature genetics (1995-09-01)
P P Pandolfi, M E Roth, A Karis, M W Leonard, E Dzierzak, F G Grosveld, J D Engel, M H Lindenbaum
ABSTRACT
GATA-3 is one member of a growing family of related transcription factors which share a strongly conserved expression pattern in all vertebrate organisms. In order to elucidate GATA-3 function using a direct genetic approach, we have disrupted the murine gene by homologous recombination in embryonic stem cells. Mice heterozygous for the GATA3 mutation are fertile and appear in all respects to be normal, whereas homozygous mutant embryos die between days 11 and 12 postcoitum (p.c.) and display massive internal bleeding, marked growth retardation, severe deformities of the brain and spinal cord, and gross aberrations in fetal liver haematopoiesis.