MilliporeSigma
  • In vitro endocrine disruption potential of organophosphate flame retardants via human nuclear receptors.
In vitro endocrine disruption potential of organophosphate flame retardants via human nuclear receptors.
Toxicology (2013-09-21)
Hiroyuki Kojima, Shinji Takeuchi, Toshihiro Itoh, Mitsuru Iida, Satoshi Kobayashi, Takahiko Yoshida
ABSTRACT
Various organophosphate flame retardants (OPFRs) are widely used in building materials, textiles and electric appliances, and have been reported to cause indoor environmental pollution in houses and office buildings. In this study, using cell-based transactivation assays, we characterized the agonistic and/or antagonistic activities of 11 OPFRs against human nuclear receptors; estrogen receptor α (ERα), ERβ, androgen receptor (AR), glucocorticoid receptor (GR), thyroid hormone receptor α1 (TRα1), TRβ1, retinoic acid receptor α (RARα), retinoid X receptor α (RXRα), pregnane X receptor (PXR), peroxisome proliferator-activated receptor α (PPARα), and PPARγ. Of the 11 OPFRs tested, triphenyl phosphate (TPhP) and tricrecyl phosphate (TCP) showed ERα and/or ERβ agonistic activity. In addition, tributyl phosphate (TBP), tris(1,3-dichloro-2-propyl) phosphate (TDCPP), TPhP and TCP showed AR antagonistic activity, and TBP, tris(2-ethylhexyl) phosphate (TEHP), TDCPP, TPhP and TCP showed GR antagonistic activity. Furthermore, we found that seven compounds, TBP, tris(2-chloro-1-methylethyl) phosphate (TCPP), TEHP, tris(2-butoxyethyl) phosphate (TBEP), TDCPP, TPhP, and TCP, display PXR agonistic activity. However, none of test compounds showed agonistic or antagonistic activity against TRα/β, or agonistic activity against RARα, RXRα or PPARα/γ. Taken together, these results suggest that several OPFRs may have potential endocrine disrupting effects via ERα, ERβ, AR, GR and PXR.
Sigma-Aldrich
Tris(2-butoxyethyl) phosphate, 94%
Pricing and availability is not currently available.
Sigma-Aldrich
Tris(2-ethylhexyl) phosphate, 97%
Pricing and availability is not currently available.