MilliporeSigma
  • Purification, crystallization and preliminary X-ray investigation of quinoprotein methylamine dehydrogenase from Thiobacillus versutus.
Purification, crystallization and preliminary X-ray investigation of quinoprotein methylamine dehydrogenase from Thiobacillus versutus.
European journal of biochemistry (1986-01-15)
F M Vellieux, J Frank, M B Swarte, H Groendijk, J A Duine, J Drenth, W G Hol
ABSTRACT
The enzyme methylamine dehydrogenase or primary-amine:(acceptor) oxidoreductase (deaminating) (EC 1.4.99.3) was purified from the bacterium Thiobacillus versutus to homogeneity, as judged by polyacrylamide gel electrophoresis. The native enzyme has a Mr of 123 500 and contains four subunits arranged in a alpha 2 beta 2 configuration, the light and heavy subunits having a Mr of 12900 and 47500 respectively. The isoelectric point is 3.9. The purified enzyme was crystallized from 37--42% saturated ammonium sulphate in 0.1 M sodium acetate buffer, pH 5.0. The space group is P3(1)21 or P3(2)21, with one alpha 2 beta 2 molecule in the asymmetric unit. The cell dimensions are: a = b = 13.01 nm; c = 10.40 nm. The X-ray diffraction pattern extends to at least 0.25-nm resolution.
Sigma-Aldrich
Phosphorylase b from rabbit muscle, lyophilized powder, ≥20 units/mg protein, 2× crystallization