MilliporeSigma
  • Self-assembly in surfactant-based liquid mixtures: bis(2-ethylhexyl)phosphoric acid/bis(2-ethylhexyl)amine systems.
Self-assembly in surfactant-based liquid mixtures: bis(2-ethylhexyl)phosphoric acid/bis(2-ethylhexyl)amine systems.
Journal of colloid and interface science (2010-05-15)
Pietro Calandra, Angela Ruggirello, Andrea Mele, Vincenzo Turco Liveri
ABSTRACT
Surfactant-based liquid mixtures constitute an interesting class of nanostructured materials with promising potential in specialized applications. Here, structural and conductometric properties of liquid mixtures composed of bis(2-ethylhexyl)amine (BEEA) and bis(2-ethylhexyl)phosphoric acid (HDEHP) have been thoroughly investigated with the aim to correlate structural features with system charge transport capability. The evolution of self-assembled local nanostructures with system composition has been investigated by FT-IR and XRD while the conductometric properties were probed by conventional AC complex impedance. Both pure components exhibit nano-segregation due to their amphiphilic nature but with only very low proton conductivities. However their mixtures, characterized by local organization of polar and apolar domains driven by acid-base interactions between HDEHP PO(4)H and BEEA NH groups, show enhanced structural order and proton conductivity.