MilliporeSigma
  • Increased open circuit voltage in fluorinated benzothiadiazole-based alternating conjugated polymers.
Increased open circuit voltage in fluorinated benzothiadiazole-based alternating conjugated polymers.
Chemical communications (Cambridge, England) (2011-09-13)
Yong Zhang, Shang-Chieh Chien, Kung-Shih Chen, Hin-Lap Yip, Ying Sun, Joshua A Davies, Fang-Chung Chen, Alex K-Y Jen
ABSTRACT
Small band-gap conjugated polymers based on monofluoro- and difluoro-substituted benzothiadiazole were developed. Highly efficient polymer solar cells (PCE as high as 5.40%) could be achieved for devices made from these polymers.