- Highly soluble [1]benzothieno[3,2-b]benzothiophene (BTBT) derivatives for high-performance, solution-processed organic field-effect transistors.
Highly soluble [1]benzothieno[3,2-b]benzothiophene (BTBT) derivatives for high-performance, solution-processed organic field-effect transistors.
2,7-Dialkyl[1]benzothieno[3,2-b]benzothiophenes were tested as solution-processible molecular semiconductors. Thin films of the organic semiconductors deposited on Si/SiO2 substrates by spin coating have well-ordered structures as confirmed by XRD analysis. Evaluations of the devices under ambient conditions showed typical p-channel FET responses with the field-effect mobility higher than 1.0 cm2 V-1 s-1 and Ion/Ioff of approximately 10(7).