MilliporeSigma
  • A novel function for fatty acid translocase (FAT)/CD36: involvement in long chain fatty acid transfer into the mitochondria.
A novel function for fatty acid translocase (FAT)/CD36: involvement in long chain fatty acid transfer into the mitochondria.
The Journal of biological chemistry (2004-05-27)
Shannon E Campbell, Narendra N Tandon, Gebretateos Woldegiorgis, Joost J F P Luiken, Jan F C Glatz, Arend Bonen
ABSTRACT
Fatty acid translocase (FAT)/CD36 is a long chain fatty acid transporter present at the plasma membrane, as well as in intracellular pools of skeletal muscle. In this study, we assessed the unexpected presence of FAT/CD36 in both subsarcolemmal and intermyofibril fractions of highly purified mitochondria. Functional assessments demonstrated that the mitochondria could bind (14)C-labeled palmitate, but could only oxidize it in the presence of carnitine. However, the addition of sulfo-N-succinimidyl oleate, a known inhibitor of FAT/CD36, resulted in an 87 and 85% reduction of palmitate oxidation in subsarcolemmal and intermyofibril fractions, respectively. Further studies revealed that maximal carnitine palmitoyltransferase I (CPTI) activity in vitro was inhibited by succinimidyl oleate (42 and 48% reduction). Interestingly, CPTI immunoprecipitated with FAT/CD36, indicating a physical pairing. Tissue differences in mitochondrial FAT/CD36 protein follow the same pattern as the capacity for fatty acid oxidation (heart > red muscle > white muscle). Additionally, chronic stimulation of hindlimb muscles (7 days) increased FAT/CD36 expression and also resulted in a concomitant increase in mitochondrial FAT/CD36 content (46 and 47% increase). Interestingly, with acute electrical stimulation of hindlimb muscles (30 min), FAT/CD36 expression was not altered, but there was an increase in the mitochondrial content of FAT/CD36 compared with the non-stimulated control limb (35 and 37% increase). Together, these data suggest a role for FAT/CD36 in mitochondrial long chain fatty acid uptake and demonstrate system flexibility to match FAT/CD36 mitochondrial content with an increased capacity for fatty acid oxidation, possibly involving translocation of FAT/CD36 to the mitochondria.
Sigma-Aldrich
Protease from Bacillus licheniformis, Type VIII, lyophilized powder, 7-15 units/mg solid