MilliporeSigma
  • Anti-Candida properties of asaronaldehyde of Acorus gramineus rhizome and three structural isomers.
Anti-Candida properties of asaronaldehyde of Acorus gramineus rhizome and three structural isomers.
Chinese medicine (2013-09-10)
Sandeep B Rajput, Ravikumar B Shinde, Madhushree M Routh, Sankunny M Karuppayil
ABSTRACT
Asaronaldehyde (2, 4, 5-trimethoxybeznaldehyde) is an active component of Acorus gramineus rhizome. This study aims to evaluate the anti-Candida efficacy of asaronaldehyde and its three structural isomers, namely, 2, 3, 4-trimethoxybenzaldehyde, 3, 4, 5-trimethoxybenzaldehyde, and 2, 4, 6- trimethoxybenzaldehyde. Susceptibility testing of test compounds was carried out using standard methodology (M27-A2) as per clinical and laboratory standards institute guidelines. Minimum fungicidal concentration (MFC) was determined as the lowest concentration of drug killing 99.9% of Candida cells. The effect on sterol profile was evaluated using the ergosterol quantitation method. Effects on morphogenesis, adhesion and biofilm formation in C. albicans were studied using germ-tube, adherence and biofilm formation assays respectively. Cytotoxicity of test compounds to human RBCs was determined by hemolysis assay. 2, 4, 6-Trimethoxybenzaldehyde exhibited significant anti-Candida activity (P = 0.0412). Minimum inhibitory concentration (MIC) and minimum fungicidal concentration (MFC) were established as 0.25 and 0.5 mg/mL, respectively. All of the test compounds showed significant inhibition of hyphal form transition in yeast at MIC/2 and MIC/4 values. 3, 4, 5-Trimethoxybenzaldehyde and 2, 4, 6-trimethoxybenzaldehyde inhibited adhesion and biofilms. A hemolytic assay of these compounds revealed that they were non-toxic at MIC values. Asaronaldehyde reduced sterol content. Asaronaldehyde and 2, 4, 6-trimethoxybenzaldehyde showed anti-Candida efficacy.
Sigma-Aldrich
2,4,6-Trimethoxybenzaldehyde, 98%
Pricing and availability is not currently available.