MilliporeSigma
Cardiomyocyte-transistor-hybrids for sensor application.
Biosensors & bioelectronics (2001-09-07)
S Ingebrandt, C K Yeung, M Krause, A Offenhäusser
ABSTRACT
An extracellular recording system has been designed for the detection of electrical cell signals using p-channel or n-channel field-effect transistor (FET) arrays with non-metallized gates. Signals from rat heart muscle cell were recorded by these devices and the results described on the basis of an equivalent circuit. This technique is sensitive enough to detect minute changes of the extracellular membrane voltage and has potential applications in drug screening. We show that known cardiac stimulants (isoproterenol, norepinephrine) and relaxants (verapamil, carbamylcholine) have characteristic effects on the heart cells in terms of the changes of beat frequencies in the absence or presence of corresponding agents.
Sigma-Aldrich
Deoxyribonuclease II from bovine spleen, Type V, essentially salt-free, lyophilized powder, ≥1,000 units/mg protein
Pricing and availability is not currently available.