MilliporeSigma
  • Distinct isoforms of the Drosophila Brd4 homologue are present at enhancers, promoters and insulator sites.
Distinct isoforms of the Drosophila Brd4 homologue are present at enhancers, promoters and insulator sites.
Nucleic acids research (2013-08-16)
Wendy A Kellner, Kevin Van Bortle, Li Li, Edward Ramos, Naomi Takenaka, Victor G Corces
ABSTRACT
Brd4 is a double bromodomain protein that has been shown to interact with acetylated histones to regulate transcription by recruiting Positive Transcription Elongation Factor b to the promoter region. Brd4 is also involved in gene bookmarking during mitosis and is a therapeutic target for the treatment of acute myeloid leukemia. The Drosophila melanogaster Brd4 homologue is called Fs(1)h and, like its vertebrate counterpart, encodes different isoforms. We have used ChIP-seq to examine the genome-wide distribution of Fs(1)h isoforms. We are able to distinguish the Fs(1)h-L and Fs(1)h-S binding profiles and discriminate between the genomic locations of the two isoforms. Fs(1)h-S is present at enhancers and promoters and its amount parallels transcription levels. Correlations between the distribution of Fs(1)h-S and various forms of acetylated histones H3 and H4 suggest a preference for binding to H3K9acS10ph. Surprisingly, Fs(1)h-L is located at sites in the genome where multiple insulator proteins are also present. The results suggest that Fs(1)h-S may be responsible for the classical role assigned to this protein, whereas Fs(1)h-L may have a new and unexpected role in chromatin architecture by working in conjunction with insulator proteins to mediate intra- or inter-chromosome interactions.
Sigma-Aldrich
Deoxyribonuclease I bovine, recombinant, expressed in Pichia pastoris, buffered aqueous glycerol solution, ≥5,000 units/mg protein