MilliporeSigma
  • DG3173 (somatoprim), a unique somatostatin receptor subtypes 2-, 4- and 5-selective analogue, effectively reduces GH secretion in human GH-secreting pituitary adenomas even in Octreotide non-responsive tumours.
DG3173 (somatoprim), a unique somatostatin receptor subtypes 2-, 4- and 5-selective analogue, effectively reduces GH secretion in human GH-secreting pituitary adenomas even in Octreotide non-responsive tumours.
European journal of endocrinology (2011-11-09)
U Plöckinger, U Hoffmann, M Geese, A Lupp, M Buchfelder, J Flitsch, P Vajkoczy, W Jakob, W Saeger, S Schulz, C Dohrmann
ABSTRACT
Somatostatin analogues (SSA) reduce autonomous GH secretion by activating somatostatin receptors (sst) 2 and 5 in 50-60% of acromegalic patients. However, by inhibiting insulin secretion these SSA reduce glucose tolerance. DG3173 is a novel SSA with additional binding to sst4 and low insulin-suppressing activity. We investigated the effect of DG3173, including its relation to specific tumour characteristics, on GH secretion in human somatotroph adenoma cell cultures (hSA) in comparison with Octreotide. Twenty-seven hSA were characterised immunohistochemically for their hormone- and sst-expression, granularity and pre-surgical therapy with SSA. GH was determined in supernatants of hSA treated with DG3173 or Octreotide in time- (n=6) and dose-response (n=21) experiments. A positive response was defined as GH suppression to below 80% of baseline. In the dose-response experiments DG3173 suppressed GH secretion in more adenomas than Octreotide (10/21 vs 5/21), including 38% (6/16) of Octreotide non-responders. In responders the extent of GH suppression and IC(50) were comparable for both SSA. The response-rate of both SSA was higher in monohormonal vs bihormonal adenomas, yet GH declined similarly in both groups. Neither pre-surgical SSA (n=6) nor tumour morphology was related to the GH response. However, semi-quantitative analysis indicated a small but significant negative correlation between the GH response to Octreotide and the immunoreactivity scores of sst2 expression. DG3173 equalled Octreotide in suppressing GH secretion in hSA. Since DG3173 suppressed GH in some Octreotide-non-responsive adenomas, its clinical effectiveness will be worth testing. Moreover, its reduced insulin-suppressive potency would make it a valuable alternative to Octreotide.
Sigma-Aldrich
Trypsin inhibitor from Glycine max (soybean), powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Deoxyribonuclease II from porcine spleen, Type IV, lyophilized powder, 2,000-6,000 Kunitz units/mg protein (biuret)
Pricing and availability is not currently available.