MilliporeSigma
Get up to 22% off for Pi Day until 3/26.Save Now
  • Membrane on a chip: a functional tethered lipid bilayer membrane on silicon oxide surfaces.
Membrane on a chip: a functional tethered lipid bilayer membrane on silicon oxide surfaces.
Biophysical journal (2005-08-30)
Vladimir Atanasov, Nikolaus Knorr, Randolph S Duran, Sven Ingebrandt, Andreas Offenhäusser, Wolfgang Knoll, Ingo Köper
ABSTRACT
Tethered membranes have been proven during recent years to be a powerful and flexible biomimetic platform. We reported in a previous article on the design of a new architecture based on the self-assembly of a thiolipid on ultrasmooth gold substrates, which shows extremely good electrical sealing properties as well as functionality of a bilayer membrane. Here, we describe the synthesis of lipids for a more modular design and the adaptation of the linker part to silane chemistry. We were able to form a functional tethered bilayer lipid membrane with good electrical sealing properties covering a silicon oxide surface. We demonstrate the functional incorporation of the ion carrier valinomycin and of the ion channel gramicidin.
Sigma-Aldrich
(±)-3-Benzyloxy-1,2-propanediol, ≥97.0% (HPLC)