MilliporeSigma
  • Difluoromethyl 2-pyridyl sulfone: a new gem-difluoroolefination reagent for aldehydes and ketones.
Difluoromethyl 2-pyridyl sulfone: a new gem-difluoroolefination reagent for aldehydes and ketones.
Organic letters (2010-03-10)
Yanchuan Zhao, Weizhou Huang, Lingui Zhu, Jinbo Hu
ABSTRACT
Difluoromethyl 2-pyridyl sulfone, a previously unknown compound, was found to act as a novel and efficient gem-difluoroolefination reagent for both aldehydes and ketones. It was found that the fluorinated sulfinate intermediate in the reaction is relatively stable, which can be observed by (19)F NMR and trapped with CH(3)I.
Sigma-Aldrich
Difluoromethyl 2-pyridyl sulfone, 97% (HPLC)