MilliporeSigma
  • Synthesis of the highly selective p38 MAPK inhibitor UR-13756 for possible therapeutic use in Werner syndrome.
Synthesis of the highly selective p38 MAPK inhibitor UR-13756 for possible therapeutic use in Werner syndrome.
Future medicinal chemistry (2011-03-24)
Mark C Bagley, Terence Davis, Michal J Rokicki, Caroline S Widdowson, David Kipling
ABSTRACT
UR-13756 is a potent and selective p38 mitogen-activated protein kinase (MAPK) inhibitor, reported to have good bioavailability and pharmacokinetic properties and, thus, is of potential use in the treatment of accelerated aging in Werner syndrome. Irradiation of 2-chloroacrylonitrile and methylhydrazine in ethanol at 100 °C gives 1-methyl-3-aminopyrazole, which reacts with 4-fluorobenzaldehyde and a ketone, obtained by Claisen condensation of 4-picoline, in a Hantzsch-type 3-component hereocyclocondensation, to give the pyrazolopyridine UR-13756. UR-13756 shows p38 MAPK inhibitory activity in human telomerase reverse transcriptase-immortalized HCA2 dermal fibroblasts, with an IC(50) of 80 nm, as shown by ELISA, is 100% efficacious for up to 24 h at 1.0 μm and displays excellent kinase selectivity over the related stress-activated c-Jun kinases. In addition, UR-13756 is an effective p38 inhibitor at 1.0 μm in Werner syndrome cells, as shown by immunoblot. The convergent synthesis of UR-13756 is realized using microwave dielectric heating and provides a highly selective inhibitor that shows excellent selectivity for p38 MAPK over c-Jun N-terminal kinase.
Sigma-Aldrich
4-Fluorobenzaldehyde, 98%
Pricing and availability is not currently available.