MilliporeSigma
A family of Acrp30/adiponectin structural and functional paralogs.
Proceedings of the National Academy of Sciences of the United States of America (2004-07-03)
Guang W Wong, Jin Wang, Christopher Hug, Tsu-Shuen Tsao, Harvey F Lodish
ABSTRACT
Biochemical, genetic, and animal studies in recent years have established a critical role for the adipokine Acrp30/adiponectin in controlling whole-body metabolism, particularly by enhancing insulin sensitivity in muscle and liver, and by increasing fatty acid oxidation in muscle. We describe a widely expressed and highly conserved family of adiponectin paralogs designated as C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. In the present study, we focus on mCTRP2, the mouse paralog most similar to adiponectin. At nanomolar concentrations, bacterially produced mCTRP2 rapidly induced phosphorylation of AMP-activated protein kinase, acetyl-CoA carboxylase, and mitogen-activated protein kinase in C2C12 myotubes, which resulted in increased glycogen accumulation and fatty acid oxidation. The discovery of a family of adiponectin paralogs has implications for understanding the control of energy homeostasis and could provide new targets for pharmacologic intervention in metabolic diseases such as diabetes and obesity.
Product Description
Roche
Amyloglucosidase, from Aspergillus niger