MilliporeSigma
  • CARD11 and CARD14 are novel caspase recruitment domain (CARD)/membrane-associated guanylate kinase (MAGUK) family members that interact with BCL10 and activate NF-kappa B.
CARD11 and CARD14 are novel caspase recruitment domain (CARD)/membrane-associated guanylate kinase (MAGUK) family members that interact with BCL10 and activate NF-kappa B.
The Journal of biological chemistry (2001-03-30)
J Bertin, L Wang, Y Guo, M D Jacobson, J L Poyet, S M Srinivasula, S Merriam, P S DiStefano, E S Alnemri
ABSTRACT
The caspase recruitment domain (CARD) is a protein-binding module that mediates the assembly of CARD-containing proteins into apoptosis and NF-kappaB signaling complexes. We report here that CARD protein 11 (CARD11) and CARD protein 14 (CARD14) are novel CARD-containing proteins that belong to the membrane-associated guanylate kinase (MAGUK) family, a class of proteins that functions as molecular scaffolds for the assembly of multiprotein complexes at specialized regions of the plasma membrane. CARD11 and CARD14 have homologous structures consisting of an N-terminal CARD domain, a central coiled-coil domain, and a C-terminal tripartite domain comprised of a PDZ domain, an Src homology 3 domain, and a GUK domain with homology to guanylate kinase. The CARD domains of both CARD11 and CARD14 associate specifically with the CARD domain of BCL10, a signaling protein that activates NF-kappaB through the IkappaB kinase complex in response to upstream stimuli. When expressed in cells, CARD11 and CARD14 activate NF-kappaB and induce the phosphorylation of BCL10. These findings suggest that CARD11 and CARD14 are novel MAGUK family members that function as upstream activators of BCL10 and NF-kappaB signaling.