MilliporeSigma
  • Human growth differentiation factor 9 (GDF-9) and its novel homolog GDF-9B are expressed in oocytes during early folliculogenesis.
Human growth differentiation factor 9 (GDF-9) and its novel homolog GDF-9B are expressed in oocytes during early folliculogenesis.
The Journal of clinical endocrinology and metabolism (1999-08-12)
J Aaltonen, M P Laitinen, K Vuojolainen, R Jaatinen, N Horelli-Kuitunen, L Seppä, H Louhio, T Tuuri, J Sjöberg, R Bützow, O Hovata, L Dale, O Ritvos
ABSTRACT
Growth differentiation factor 9 (GDF-9) is a transforming growth factor-beta family member that is required for normal folliculogenesis in female mice, but its role as a regulator of human fertility is still unclear. We determined here by in situ hybridization and immunohistochemical analyses the localization of the GDF-9 messenger ribonucleic acid (mRNA) and protein during human folliculogenesis. The GDF-9 transcripts were not detected in primordial follicles, but they are abundantly expressed in primary follicles in frozen sections of ovarian cortical tissue material obtained at laparoscopic surgery. We raised antipeptide antibodies against GDF-9 and showed by immunohistochemical studies on paraffin sections of whole human ovaries that the GDF-9 protein is most abundantly expressed in primary follicles. We recently demonstrated that a novel GDF-9-related factor, GDF-9B, is coexpressed with GDF-9 during murine folliculogenesis. We now isolated human GDF-9B complementary DNA and genomic clones and report the unusually restricted expression pattern of human GDF-9B. The human GDF-9B transcript can be detected only in the gonads by RT-PCR analysis, and in situ hybridization studies indicate that it is not expressed in small primary follicles but, rather, in the oocytes of late primary follicles. Functional studies using the Xenopus laeuis embryo model indicate that unlike the transforming growth factor-beta family members activin and bone morphogenetic protein-4, neither GDF-9 nor GDF-9B affects mesoderm induction, suggesting that they may use signaling pathways distinct from those well defined for activin and bone morphogenetic protein-4. We conclude that 1) both GDF-9 mRNA and protein are abundantly expressed in oocytes of primary follicles in human ovary, suggesting that the GDF-9 transcript is translated at this early stage of folliculogenesis; 2) human GDF-9B is specifically expressed in gonads at low levels; and 3) the expression of GDF-9 mRNA begins slightly earlier than that of GDF-9B in the human oocytes during follicular development. Our results are consistent with the suggestion that GDF-9 and GDF-9B may regulate human folliculogenesis in a manner specific to the ovary.
Sigma-Aldrich
Anti-BMP15 antibody produced in rabbit, affinity isolated antibody