MilliporeSigma
  • Interaction of 5-amino-1,3,4-thiadiazole-2-thiol and its violuric acid adduct with Pt(II) - crystals structures, spectroscopic properties and cytotoxic activity.
Interaction of 5-amino-1,3,4-thiadiazole-2-thiol and its violuric acid adduct with Pt(II) - crystals structures, spectroscopic properties and cytotoxic activity.
Arzneimittel-Forschung (2012-10-25)
M Kavlakova, A Bakalova, G Momekov, D Ivanov
ABSTRACT
The coordination properties of Pt(II) with 5-amino-1,3,4-thiadiazole-2-thiol [CAS 2349-67-9] (L 1 ) and its novel violurate adduct (L 2 ), both in solution and in solid state, are studied by means of conventional IR-spectroscopy, single crystal X-ray diffraction and thermal methods. The complex compounds of L 1 and L 2 , with general formulas [Pt(C2H2N3S2)2] and [Pt(C6H4N6S2O3)(Cl)]Cl respectively, are obtained. Quantum chemical calculations of the ligands are performed with a view to obtain electronic structure and optical properties of the ligands L 1 and L 2 , respectively. The cell viability of the ligands and metal complexes on a panel of human tumor cell lines is evaluated.