MilliporeSigma
  • Mutations in the immunoglobulin-like domain of gp190, the leukemia inhibitory factor (LIF) receptor, increase or decrease its affinity for LIF.
Mutations in the immunoglobulin-like domain of gp190, the leukemia inhibitory factor (LIF) receptor, increase or decrease its affinity for LIF.
The Journal of biological chemistry (2003-02-26)
Juliette Bitard, Sophie Daburon, Laurence Duplomb, Frédéric Blanchard, Patricia Vuisio, Yannick Jacques, Anne Godard, John K Heath, Jean-François Moreau, Jean-Luc Taupin
ABSTRACT
The leukemia inhibitory factor (LIF) receptor comprises the low affinity binding chain gp190 and the high affinity converter gp130. The ectodomain of gp190 is among the most complex in the hematopoietin receptor family, because it contains two typical cytokine receptor homology domains separated by an immunoglobulin-like (Ig-like) domain. Human and murine gp190 proteins share 76% homology, but murine gp190 binds human LIF with a much higher affinity, a property attributed to the Ig-like domain. Using alanine-scanning mutagenesis of the Ig-like domain, we mapped a LIF binding site at its carboxyl terminus, mainly involving residue Phe-328. Mutation of selected residues into their orthologs in the murine receptor (Q251E and N321D) significantly increased the affinity for human LIF. Interestingly, these residues, although localized at both the amino and carboxyl terminus, make a spatially unique LIF binding site in a structural model of the Ig-like module. These results demonstrate definitively the role of the Ig-like domain in LIF binding and the potential to modulate receptor affinity in this family with very limited amino acid changes.