MilliporeSigma
  • Efficient asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-alpha-methylcysteine using camphorsultam as a chiral auxiliary.
Efficient asymmetric synthesis of (S)- and (R)-N-Fmoc-S-trityl-alpha-methylcysteine using camphorsultam as a chiral auxiliary.
The Journal of organic chemistry (2004-06-19)
Satendra Singh, Samala J Rao, Michael W Pennington
ABSTRACT
(1R)-(+)-2,10- and (1S)-(-)-2,10-camphorsultam were acylated with ethyl 2-phenylthiazoline 4-carboxylate to afford (+)- and (-)-2-phenylthiazolinylcamphorsultam, which were stereoselectively alkylated with MeI in the presence of n-BuLi. Alkylation of these phenylthiazolinylcamphorsultams occurred from the beta-face rather than alpha-face, resulting in the formation of (S)-alpha-methylcysteine from (1R)-(+)-2,10-camphorsultam and (R)-alpha-methylcysteine from (1S)-(-)-2,10-camphorsultam after acidic hydrolysis. Subsequent protection of the side chain thiol group with trityl alcohol and alpha-amine function with Fmoc-OSu furnished fully protected (S)- and (R)-N-Fmoc-S-trityl-alpha-methylcysteine in overall 20% yield.
Sigma-Aldrich
Ethyl benzimidate hydrochloride, ≥97.0% (AT)
Sigma-Aldrich
(1S)-(−)-2,10-Camphorsultam, 98%