MilliporeSigma
  • Palladium(II) acetate mediated oxidative cyclization of ω-unsaturated α-cyano ketones for facile construction of methylenecyclohexane ring system.
Palladium(II) acetate mediated oxidative cyclization of ω-unsaturated α-cyano ketones for facile construction of methylenecyclohexane ring system.
Organic & biomolecular chemistry (2012-05-15)
Min-Tsang Hsieh, Kak-Shan Shia, Hsing-Jang Liu, Sheng-Chu Kuo
ABSTRACT
A highly efficient annulative approach towards the construction of the structurally attractive methylenecyclohexane ring was developed through a convenient 1,4-addition of 4-pentenylmagnesium bromide to 2-cyano-2-cycloalkenones followed by a Pd(II)-mediated oxidative cyclization of the resulting ω-unsaturated α-cyano ketones. Based on this newly developed protocol, polycyclic adducts bearing various ring sizes and substitutions can be prepared in moderate to high yields.
Sigma-Aldrich
Methylenecyclohexane, 98%