MilliporeSigma
  • Occurrence of tetra- and pentasaccharides with the sialyl-Le(a) structure in human milk.
Occurrence of tetra- and pentasaccharides with the sialyl-Le(a) structure in human milk.
The Journal of biological chemistry (1990-03-25)
H Kitagawa, H Nakada, Y Numata, A Kurosaka, S Fukui, I Funakoshi, T Kawasaki, I Shimada, F Inagaki, I Yamashina
ABSTRACT
The occurrence of two novel oligosaccharides in human milk was investigated. These oligosaccharides were purified by affinity chromatography on a column of an immobilized monoclonal antibody, MSW 113. Structural studies, involving 500-MHz 1H NMR spectroscopy and fast atom bombardment-mass spectrometry, indicated the structures of these compounds to be NeuAc alpha 2----3Gal beta 1----3(Fuc alpha 1----4) GlcNAc and NeuAc alpha 2----3Gal beta 1----3(Fuc alpha 1----4) GlcNac beta 1----3Gal. This constitutes the first evidence for the occurrence of N-acetylglucosamine or galactose as the reducing-end residue of human milk oligosaccharides. These two oligosaccharides bound MSW 113 to nearly the same extent as sialyl-Lea hexasaccharide but to another sialyl-Le(a) structure-directed monoclonal antibody, NS-19-9, only weakly.