MilliporeSigma
  • Characterization of adenosine receptors evoking excitation of mesenteric afferents in the rat.
Characterization of adenosine receptors evoking excitation of mesenteric afferents in the rat.
British journal of pharmacology (1998-12-24)
A J Kirkup, C Eastwood, D Grundy, I P Chessell, P P Humphrey
ABSTRACT
We examined the effects of adenosine receptor agonists and antagonists on the discharge of mesenteric afferent nerves supplying the jejunum in pentobarbitone sodium-anaesthetized rats. Adenosine (0.03-10 mg kg(-1), i.v.), NECA (0.3-300 microg kg(-1), i.v.) and the A1 receptor agonist, GR79236 (0.3-1000 microg kg(-1), i.v.), each induced dose-dependent increases in afferent nerve activity and intrajejunal pressure, hypotension and bradycardia. The A1 receptor antagonist, DPCPX (3 mg kg(-1), i.v.), antagonized all the effects of GR79236 but only the haemodynamic effects of adenosine and NECA. The A2A receptor antagonist, ZM241385 (3 mg kg(-1), i.v.), antagonized the hypotensive effect of NECA but none of the effects of GR79236. The A2A receptor agonist, CGS21680 (0.3-300 microg kg(-1), i.v.), and the A3 receptor agonist, IB-MECA (0.3-300 microg kg(-1), i.v.), each induced only a dose-dependent hypotension. Subsequent administration of adenosine (3 mg kg(-1), i.v.) induced increases in afferent nerve activity and intrajejunal pressure and bradycardia. ZM241385 (3 mg kg(-1), i.v.) antagonized the hypotensive effect of CGS21680 but not the effects of adenosine. Bethanechol (300 microg kg(-1), i.v.) evoked increases in afferent nerve activity and intrajejunal pressure, hypotension and bradycardia. However, adenosine (3 mg kg(-1), i.v.) evoked greater increases in afferent nerve activity than bethanechol despite inducing smaller increases in intrajejunal pressure. In summary, A1 and A2B and/or A2B-like receptors evoke adenosine-induced increases in mesenteric afferent nerve activity and intrajejunal pressure in the anaesthetized rat. Furthermore, elevations in intrajejunal pressure do not wholly account for adenosine-evoked excitation of mesenteric afferent nerves.