MilliporeSigma
  • Nitric oxide synthesis by tracheal smooth muscle cells by a nitric oxide synthase-independent pathway.
Nitric oxide synthesis by tracheal smooth muscle cells by a nitric oxide synthase-independent pathway.
The American journal of physiology (1998-11-14)
Y Jia, M Zacour, B Tolloczko, J G Martin
ABSTRACT
Nitric oxide (NO) is known to be synthesized from L-arginine in a reaction catalyzed by NO synthase. Liver cytochrome P-450 enzymes also catalyze the oxidative cleavage of C==N bonds of compounds containing a -C(NH2)==NOH function, producing NO in vitro. The present study was designed to investigate whether there was evidence of a similar pathway for the production of NO in tracheal smooth muscle cells. Formamidoxime (10(-2) to 10(-4) M), a compound containing -C(NH2)==NOH, relaxed carbachol-contracted tracheal rings and increased intracellular cGMP in cultured tracheal smooth muscle cells, whereas L-arginine had no such effect. NO was detectable in the medium containing cultured tracheal smooth muscle cells when incubated with formamidoxime. Ethoxyresorufin (10(-7) to 10(-4) M), an alternate cytochrome P-450 substrate, inhibited formamidoxime-induced cGMP accumulation as well as tracheal ring relaxation in cultured tracheal smooth muscle cells. The NO synthase inhibitors Nomega-nitro-L-arginine (10(-3) M) and NG-monomethyl-L-arginine (10(-3) M) had no effect on formamidoxime-induced cGMP accumulation. These results suggest that NO can be synthesized from formamidoxime in tracheal smooth muscle cells, presumably by a reaction catalyzed by cytochrome P-450.