MilliporeSigma
  • Hydrogen peroxide-supported activities of semisynthetic flavocytochrome 2B4.
Hydrogen peroxide-supported activities of semisynthetic flavocytochrome 2B4.
Biochemistry and molecular biology international (1998-06-23)
V V Shumyantseva, Y L Avdeenko, T L Moskvitina, T V Bulko, A I Archakov
ABSTRACT
Semisynthetic flavocytochromes, obtained by covalent binding of riboflavin with cytochromes P450 2B4, were able to catalyze the H2O2-mediated reactions of aniline p-hydroxylation, aminopyrine N-demethylation and p-nitroanizole' O-dealkylation. The rates of the flavocytochrome-catalyzed, H2O2-supported reactions far exceeded those of the appropriate NADH-dependent reactions and were comparable with the cytochrome P450 2B4-catalyzed, peroxide-mediated reaction rates. The kinetic parameters (kcat, K(m)) for the peroxide-dependent flavocytochrome P450 2B4 reactions were obtained. Sodium cyanide and SKF-525A, a specific P450 inhibitor, were both shown to inhibit these reactions. The generation of active oxygen species by flavocytochrome 2B4 was registered by chemiluminescence intensity.
Sigma-Aldrich
4-Nitroanisole, 97%