MilliporeSigma
  • Properties of ginseng saponin inhibition of catecholamine secretion in bovine adrenal chromaffin cells.
Properties of ginseng saponin inhibition of catecholamine secretion in bovine adrenal chromaffin cells.
European journal of pharmacology (1998-05-23)
K Kudo, E Tachikawa, T Kashimoto, E Takahashi
ABSTRACT
To investigate the relationship between the inhibitory effects of ginseng saponins (ginsenosides) on acetylcholine-evoked secretion of catecholamines and the structures of ginsenosides, we examined the effects of ginsenoside-Rg3 and -Rh2, which are panaxadiol saponins, 20(R)- and 20(S)-ginsenoside-Rg2, which are epimers involving the hydroxyl group at C-20 of sapogenin, and other plant saponins on the acetylcholine-evoked secretion of catecholamines from cultured bovine adrenal chromaffin cells. The ginsenoside-Rg3 (1-100 microM) and -Rh2 (10-100 microM) greatly reduced the acetylcholine-evoked secretion in a concentration-dependent manner comparable to that of ginsenoside-Rg2, a panaxatriol saponin, which was the most potent inhibitor in our previous study. 20(R)- and 20(S)-ginsenoside-Rg2 (1-100 microM) similarly reduced the acetylcholine-evoked secretion. In contrast, saikosaponin-a, glycyrrhizin and the cardiac glycosides (100 nM-100 microM), digitoxin and digoxin, had no significant inhibitory effect on catecholamine secretion. Saikosaponin-c (10-100 microM), however, had an inhibitory effect, which was less than that of ginsenoside-Rg2 and -Rg3. These results strongly suggest that the inhibitory effects of ginsenosides on the acetylcholine-evoked secretion of catecholamines from bovine adrenal chromaffin cells are a unique property of ginseng. Further, the relationship between the inhibitory effects and the structures of ginsenosides is discussed.
Product Description
Supelco
Ginsenoside Rg2, analytical standard