MilliporeSigma
  • Effects of dopaminergic agonists and antagonists on the serum prolactin levels in alcoholized rats.
Effects of dopaminergic agonists and antagonists on the serum prolactin levels in alcoholized rats.
Polish journal of pharmacology (1997-08-01)
J Samochowiec, Z Kleinrok, E Jagiełło-Wójtowicz, J Horodnicki, H Rommelspacher, L G Schmidt
ABSTRACT
A single dose of ethanol (1 g/kg p.o.) significantly decreased, whereas higher doses of ethanol (2 or 3 g/kg p.o.) significantly increased the serum prolactin (PRL) concentration. Administration of ethanol at a dose of 2 g/kg p.o. for 4 weeks did not affect this parameter but the ethanol withdrawal syndrome caused a significant rise in the serum PRL level. Chronic studies showed that D1-dopaminergic agonist SKF 38390 (2.5 mg/kg) significantly raised serum PRL levels in rats. This effect was reversed by long-lasting treatment of rats with ethanol and ethanol withdrawal. Pimozide (1 mg/kg), D2 antagonist, increased PRL in those rats. On the other hand, D1-antagonist SCH 23390 (0.5 mg/kg) and D2-agonist PPHT (0.5 mg/kg) were without effect in rats administered ethanol for a long period of time. In rats with ethanol withdrawal syndrome, administration of D1-antagonist SCH 23390 (0.5 mg/kg) also did not affect the serum PRL concentrations. However, D1-agonist SKF 38393 (2.5 mg/kg) and D2-antagonist pimozide (1 mg/kg) increased the serum PRL level in rats with ethanol withdrawal syndrome, whereas D2 agonist PPHT (2 mg/kg) decreased PRL level in serum. Thus, the acute effect of ethanol on PRL level appears to be dose-dependent. It seems that chronic ethanol administration and its withdrawal especially affected D1 receptor.