MilliporeSigma
  • Inhibition of human cytochrome P450-catalyzed oxidations of xenobiotics and procarcinogens by synthetic organoselenium compounds.
Inhibition of human cytochrome P450-catalyzed oxidations of xenobiotics and procarcinogens by synthetic organoselenium compounds.
Cancer research (1997-11-14)
T Shimada, K El-Bayoumy, P Upadhyaya, T R Sutter, F P Guengerich, H Yamazaki
ABSTRACT
The effects of synthetic chemopreventive organoselenium compounds 1,2-, 1,3-, and 1,4-phenylenebis(methylene)selenocyanate (o-, m-, and p-XSC, respectively), benzyl selenocyanate (BSC), and dibenzyl diselenide (DDS) and inorganic sodium selenite on the oxidation of xenobiotics and procarcinogens by human cytochrome P450 (P450 or CYP) enzymes were determined in vitro. Spectral studies showed that BSC and three XSC compounds (but not sodium selenite or DDS) induced type II difference spectrum when added to the suspension of liver microsomes isolated from beta-naphthoflavone-treated rats, with m-XSC being the most potent in inducing spectral interactions with P450 enzymes; m-XSC also produced a type II spectral change with human liver microsomes. o-, m-, and p-XSC inhibited 7-ethoxyresorufin O-deethylation catalyzed by human liver microsomes when added at concentrations below 1 microM levels, but BSC and DDS were less effective. All of these compounds inhibited the oxidation of model substrates for human P450s to varying extents. We studied the effects of these compounds on the activation of procarcinogens by recombinant human CYP1A1, 1A2, and 1B1 enzymes using Salmonella typhimurium NM2009 tester strain for the detection of DNA damage. The three XSCs were found to be very potent inhibitors of metabolic activation of 3-amino-1,4-dimethyl-5H-pyrido[4,3-b]indole, 2-amino-3,5-dimethylimidazo[4,5-f]quinoline, and 2-aminoanthracene, catalyzed by CYP1A1, 1A2, and 1B1, respectively. The potency of inhibition of m-XSC on CYP1B1-dependent activation of 2-aminoanthracene was compatible to those of alpha-naphthoflavone. These inhibitory actions may, in part, account for the mechanisms responsible for cancer prevention by organoselenium compounds in laboratory animals.