MilliporeSigma
  • Mast cell tumor necrosis factor alpha production is regulated by MEK kinases.
Mast cell tumor necrosis factor alpha production is regulated by MEK kinases.
Proceedings of the National Academy of Sciences of the United States of America (1997-06-10)
T Ishizuka, N Terada, P Gerwins, E Hamelmann, A Oshiba, G R Fanger, G L Johnson, E W Gelfand
ABSTRACT
Mast cells synthesize and secrete specific cytokines and chemokines which play an important role in allergic inflammation. Aggregation of the high-affinity Fc receptor (FcepsilonRI) for immunoglobulin E (IgE) in MC/9 mouse mast cells stimulates the synthesis and secretion of tumor necrosis factor alpha (TNF-alpha). FcepsilonRI aggregation activates several sequential protein kinase pathways, leading to increased activity of extracellular signal-regulated kinases (ERKs), c-Jun amino-terminal kinases (JNKs), and the p38 mitogen-activated protein (MAP) kinase. Inhibition of ERKs with the compound PD 098059 had little effect on FcepsilonRI-stimulated TNF-alpha production. Aggregation of FcepsilonRI stimulated MEK kinase 1 (MEKK1) activity, which activates JNK kinase (JNKK), the kinase that phosphorylates and activates JNKs. Expression of activated MEKK1 (DeltaMEKK1) in MC/9 cells strongly stimulated JNK activity but only weakly stimulated p38 activity, and it induced a large activation of TNF-alpha promoter-regulated luciferase gene expression. Inhibitory mutant JNK2 expressed in MC/9 cells significantly blunted FcepsilonRI stimulation of TNF-alpha promoter-driven luciferase expression. Wortmannin, an inhibitor of phosphatidylinositol 3-kinase, diminished FcepsilonRI-mediated TNF-alpha synthesis, significantly blunted JNK activation and TNF-alpha promoter-driven luciferase expression, and only weakly inhibited p38 kinase activation. Inhibition of NFkappaB activation resulting from DeltaMEKK1 expression or FcepsilonRI stimulation did not affect TNF-alpha promoter-driven luciferase expression. Our findings define a MEKK-regulated JNK pathway activated by FcepsilonRI that regulates TNF-alpha production in mast cells.
Sigma-Aldrich
SKF-86002, ≥98% (HPLC), solid