MilliporeSigma
In vitro anticryptosporidial activity of dinitroaniline herbicides.
FEMS microbiology letters (1996-03-01)
M J Arrowood, J R Mead, L Xie, X You
ABSTRACT
Despite the evaluation of over 100 antimicrobial drugs, the diarrheal disease cryptosporidiosis has remained refractory to treatment. We report the evaluation of five dinitroaniline herbicides including trifluralin, profluralin, nitralin, pendimethalin, and fluchloralin for anticryptosporidial activity in an in vitro cultivation model of Cryptosporidium parvum. All five compounds exhibited significant anticryptosporidial activities with no corresponding evidence of toxicity. The most active compound was pendimethalin with an IC50 of 0.19 microM while nitralin was the least active with an IC50 of 4.5 microM. These compounds should be evaluated further in an animal model of cryptosporidiosis.
Product Description
Supelco
Profluralin, PESTANAL®, analytical standard
Pricing and availability is not currently available.