MilliporeSigma
  • Evaluation of C-(beta-D-galactosyl) and C-(2-deoxy-D-lyxo-hex-1-enopyranosyl) (D-galactal type) derivatives as inhibitors of beta-D-galactosidase from Escherichia coli.
Evaluation of C-(beta-D-galactosyl) and C-(2-deoxy-D-lyxo-hex-1-enopyranosyl) (D-galactal type) derivatives as inhibitors of beta-D-galactosidase from Escherichia coli.
Carbohydrate research (1996-09-23)
L Kiss, L Somsák
ABSTRACT
C-(2-Deoxy-D-lyxo-hex-1-enopyranosyl)formamide was prepared from acetylated C-(beta-D-galactopyranosyl)formamide by a radical-mediated bromination-zinc/N-methylimidazole-induced reductive elimination-Zemplén deacetylation reaction sequence. The preparation of acetylated 5-(2-deoxy-D-lyxo-hex-1-enopyranosyl)tetrazole was improved. Methyl C-(2-deoxy-D-lyxo-hex-1-enopyranosyl)formimidate was transformed by benzylamine into N-benzyl-C-(2-deoxy-D-lyxo-hex-1-enopyranosyl)formamidine and, after hydrolysis to methyl C-(2-deoxy-D-lyxo-hex-1-enopyranosyl)formate, into N-benzyl-C-(2-deoxy-D-lyxo-hex-1-enopyranosyl)formamide. A series of C-(beta-D-galactopyranosyl) and C-(2-deoxy-D-lyxo-hex-1-enopyranosyl) derivatives was comparatively investigated for E. coli beta-D-galactosidase inhibitory activity. N-Benzyl-C-(2-deoxy-D-lyxo-hex-1-enopyranosyl)formamidine was the best inhibitor and had K(i) = 6 microM (on the basis of its free base concentration, K(i) = 8.3 nM was obtained). Basicity and hydrophobicity of the aglycon proved to be more important factors for the inhibition than the conformation of the sugar ring.