MilliporeSigma
  • Synthesis and cytoprotective antiulcer activity of 2- or 4-(1H-pyrazol-1-yl)pyrimidine derivatives related to mepirizole and dulcerozine.
Synthesis and cytoprotective antiulcer activity of 2- or 4-(1H-pyrazol-1-yl)pyrimidine derivatives related to mepirizole and dulcerozine.
Chemical & pharmaceutical bulletin (1996-09-01)
M Ikeda, K Maruyama, Y Nobuhara, T Yamada, S Okabe
ABSTRACT
(1H-Pyrazol-1-yl)-, (1H-imidazol-1-yl)-, and (1H-1,2,4-triazol-1-yl)pyrimidines were prepared and evaluated for cytoprotective antiulcer activity. Among them, 4-methoxy-6-methyl-2-(1H-pyrazol-1-yl)pyrimidine (18) showed potent inhibition of the HCl-ethanol-induced and water-immersion stress-induced ulcers in rats, as well as low acute toxicity.
Product Description
Supelco
Mepirizole, analytical standard