MilliporeSigma
RAPID TECHNIQUE FOR THE ENUMERATION OF CLOSTRIDIUM PERFINGENS.
Applied microbiology (1965-07-01)
R S MARSHALL, J F STEENBERGEN, L S MCCLUNG
ABSTRACT
A new medium, Tryptone-sulfite-neomycin (TSN) agar, and an incubation procedure for the enumeration of Clostridium perfringens are described. Tolerance to neomycin, optimal growth at 46 C, and sulfite-reducing properties of C. perfringens were used as a basis for development of the medium. Comparisons were made between sulfite-polymyxin-sulfadiazine (SPS) agar and TSN agar at 37 and 46 C with C. perfringens and other organisms. These studies indicate the quantitative and selective superiority of TSN agar, incubated at 46 C, over SPS agar.
Product Description
Millipore
TSN Agar, suitable for microbiology, NutriSelect® Plus
Pricing and availability is not currently available.