MilliporeSigma
  • Inhalation experiments with mixtures of hydrocarbons. Experimental design, statistics and interpretation of kinetics and possible interactions.
Inhalation experiments with mixtures of hydrocarbons. Experimental design, statistics and interpretation of kinetics and possible interactions.
Archives of toxicology (1996-01-01)
I Eide, K Zahlsen
ABSTRACT
The paper describes experimental and statistical methods for toxicokinetic evaluation of mixtures in inhalation experiments. Synthetic mixtures of three C9 n-paraffinic, naphthenic and aromatic hydrocarbons (n-nonane, trimethylcyclohexane and trimethylbenzene, respectively) were studied in the rat after inhalation for 12h. The hydrocarbons were mixed according to principles for statistical experimental design using mixture design at four vapour levels (75, 150, 300 and 450 ppm) to support an empirical model with linear, interaction and quadratic terms (Taylor polynome). Immediately after exposure, concentrations of hydrocarbons were measured by head space gas chromatography in blood, brain, liver, kidneys and perirenal fat. Multivariate data analysis and modelling were performed with PLS (projections to latent structures). The best models were obtained after removing all interaction terms, suggesting that there were no interactions between the hydrocarbons with respect to absorption and distribution. Uptake of paraffins and particularly aromatics is best described by quadratic models, whereas the uptake of the naphthenic hydrocarbons is nearly linear. All models are good, with high correlation (r2) and prediction properties (Q2), the latter after cross validation. The concentrations of aromates in blood were high compared to the other hydrocarbons. At concentrations below 250 ppm, the naphthene reached higher concentrations in the brain compared to the paraffin and the aromate. Statistical experimental design, multivariate data analysis and modelling have proved useful for the evaluation of synthetic mixtures. The principles may also be used in the design of liquid mixtures, which may be evaporated partially or completely.
Sigma-Aldrich
1,2,4-Trimethylcyclohexane, mixture of isomers, 97%
Pricing and availability is not currently available.