MilliporeSigma
N-cubylmethyl substituted morphinoids as novel narcotic antagonists.
Bioorganic & medicinal chemistry (1996-01-01)
C Y Cheng, L W Hsin, Y P Lin, P L Tao, T T Jong
ABSTRACT
N-Cubylmethylnormorphine (1) and N-cubylmethylnoroxymorphone (2) have been synthesized and found to be more potent ligands at the mu and kappa opioid receptors than morphine and oxymorphone respectively. In the guinea-pig ileum preparation, compounds 1 and 2 were characterized as opioid mu antagonists (Ke = 68 and 16 nM, respectively). Compound 2 also showed effective kappa-antagonism (Ke = 22 nM). The narcotic antagonism activity of 1 has been confirmed by in vivo assays.