MilliporeSigma
  • Drug-protein conjugates: haptenation of 1-methyl-10 alpha-methoxydihydrolysergol and 5-bromonicotinic acid to albumin for the production of epitope-specific monoclonal antibodies against nicergoline.
Drug-protein conjugates: haptenation of 1-methyl-10 alpha-methoxydihydrolysergol and 5-bromonicotinic acid to albumin for the production of epitope-specific monoclonal antibodies against nicergoline.
Journal of pharmaceutical sciences (1995-09-01)
F Gabor, G Hamilton, F Pittner
ABSTRACT
Two types of monoclonal antibodies were used for the determination of nicergoline in biological matrices. The antibodies were prepared with the hydrolysis products 5-bromonicotinic acid and 1-methyl-10 alpha-methoxydihydrolysergol after hemisuccinoylation to haptens. The current amide bond-generating methods (mixed anhydride-, carbodiimide-, carbodiimide/sulfo-N-hydroxysuccinimide-, and dicyclohexylcarbodiimide/N-hydroxysuccinimide methods) were used in bovine serum albumin (BSA)-coupling techniques and yielded conjugates that were haptenated to varying extents. The conjugates exhibiting 23 mol of 1-methyl-10 alpha-methoxydihydrolysergol (MMD) or 41 mol of 5-bromonicotinic acid (BNA) per mole of BSA were used for both immunization of mice and for coating the wells of the microtiter plates to select hybridomas and investigate specificity of the obtained antibodies. The results of hapten-inhibition ELISA using antigen-coated wells indicate that the supernatant of MMD-specific hybridoma exhibited 50% inhibition of antibody binding at 17 +/- 2 micrograms of MMD and at 24.5 +/- 2 micrograms of nicergoline, and the BNA-specific hybridoma exhibited similar inhibition at 147 +/- 6 micrograms of BNA and 500 +/- 30 micrograms of nicergoline. A main requirement for analytical purposes is that two different types of monoclonal antibodies recognize two different epitopes on nicergoline and its main metabolite, as shown by hapten-inhibition ELISA.
Sigma-Aldrich
5-Bromopyridine-3-carboxylic acid, 98%