MilliporeSigma
  • [Comparative study of the pharmacokinetics of amitriptyline oxide and trimipramine after single administration in healthy male probands and patients with renal failure].
[Comparative study of the pharmacokinetics of amitriptyline oxide and trimipramine after single administration in healthy male probands and patients with renal failure].
Medizinische Klinik (Munich, Germany : 1983) (1993-03-15)
J Degen, E Wölke, M Seiberling, P Pintar, G Höxter, H B Steinhauer, W Fischer
ABSTRACT
The pharmacokinetics of the antidepressants amitriptyline oxide and trimipramine and their major metabolites amitriptyline, nor-triptyline and desmethyltrimipramine, were studied in twelve healthy male subjects (aged from 22 to 62 years) and twelve patients (aged from 25 to 73 years) with severe renal impairment (glomerular filtration rate < 10 ml/min). Oral single doses of 60 mg amitriptyline oxide and 50 mg trimipramine, separated by a washout period, were administered to all study participants. Blood and urine samples were collected up to 120 hours after administration. For trimipramine and desmethyltrimipramine, a new HPLC method was developed. The "Fischer Somatic and Undesired Effects Check List" was used for the assessment of adverse events. The mean plasma half-life and AUC of amitriptyline oxide and its metabolites were significantly higher in patients than in healthy adults. For trimipramine the AUC was significantly higher in patients. The plasma half-life of trimipramine was longer in patients, but statistically not significant. The maximum plasma concentrations for both drugs and metabolites were at an average distinctly higher in patients. Clearance rate of amitroptylinoxide and trimipramine also differed between the two groups. Correlating with these results a high incidence and a longer persistence (in most cases > 12 hours) and more pronounced adverse effects were noted in the patient group, whereas in volunteers adverse events were only observed up to approximately eight hours.
Product Description
Supelco
N-Desmethyltrimipramine maleate solution, 1.0 mg/mL in methanol (as free base), ampule of 1 mL, certified reference material, Cerilliant®