MilliporeSigma
  • Pathways of dephosphorylation of 1-D-myo-inositol 1,4,5-trisphosphate in GH3 pituitary tumor cells.
Pathways of dephosphorylation of 1-D-myo-inositol 1,4,5-trisphosphate in GH3 pituitary tumor cells.
Biochimica et biophysica acta (1993-07-28)
F Ruiz-Larrea, A H Drummond
ABSTRACT
Previous work in [3H]inositol-labelled GH3 pituitary tumor cells stimulated with thyrotropin-releasing hormone (TRH) reported the existence of at least ten distinct [3H]inositol-containing substances which were identified as different inositol mono-, bis- and tris-phosphate isomers [1]. Here a complete kinetic study of the dephosphorylation pathways of the second messenger Ins(1,4,5)P3 is reported in GH3 cell homogenates, identifying a new intermediate, Ins(4,5)P2, in the metabolism of the second messenger. in vitro results obtained with exogenous substrates are compared with in vivo results obtained measuring levels of the endogenous [3H]inositol-labelled isomers that participate in the dephosphorylation pathways of Ins(1,4,5)P3 in resting and TRH-stimulated GH3 cells. The effect of Li+ on the activity of the different phosphatases involved in these pathways is studied as well.