MilliporeSigma
  • [Effectiveness of glutamate in the treatment of early manifestations of occupational fluorosis].
[Effectiveness of glutamate in the treatment of early manifestations of occupational fluorosis].
Meditsina truda i promyshlennaia ekologiia (1994-01-01)
T D Grekhova, B A Katsnelśon, V M Kolmogortseva, L K Konysheva, O M Babakova
ABSTRACT
Efficiency of glutamic acid for therapy of early signs of occupational fluorosis was studied in workers engaged into cryolite production. The study proved that use of glutamic acid in occupational conditions prevents progressing of metabolic disorders. The results encourage recommendations to include glutamate into therapeutic and prophylactic nutrition of workers exposed to fluor compounds, into nutritive additions according to special recipe.
Sigma-Aldrich
Cryolite, synthetic, ≥97.0% (from F)