MilliporeSigma
Biodegradation of 2-nitrotoluene by Pseudomonas sp. strain JS42.
Applied and environmental microbiology (1994-09-01)
B E Haigler, W H Wallace, J C Spain
ABSTRACT
A strain of Pseudomonas sp. was isolated from nitrobenzene-contaminated soil and groundwater on 2-nitrotoluene as the sole source of carbon, energy, and nitrogen. Bacterial cells growing on 2-nitrotoluene released nitrite into the growth medium. The isolate also grew on 3-methylcatechol, 4-methylcatechol, and catechol. 2-Nitrotoluene, 3-methylcatechol, and catechol stimulated oxygen consumption by intact cells regardless of the growth substrate. Crude extracts from the isolate contained catechol 2,3-dioxygenase and 2-hydroxy-6-oxohepta-2,4-dienoate hydrolase activity. The results suggest that 2-nitrotoluene is subject to initial attack by a dioxygenase enzyme that forms 3-methylcatechol with concomitant release of nitrite. The 3-methylcatechol is subsequently degraded via the meta ring fission pathway.
Sigma-Aldrich
3-Nitrotoluene, 99%
Pricing and availability is not currently available.