MilliporeSigma
  • 12-Deoxyphorbol-13-O-phenylacetate 20 acetate [an agonist of protein kinase C beta 1 (PKC beta 1)] induces DNA synthesis, interleukin-2 (IL-2) production, IL-2 receptor alpha-chain (CD25) and beta-chain (CD122) expression, and translocation of PKC beta isozyme in human peripheral blood lymphocytes: evidence for a role of PKC beta 1 in human T cell activation.
12-Deoxyphorbol-13-O-phenylacetate 20 acetate [an agonist of protein kinase C beta 1 (PKC beta 1)] induces DNA synthesis, interleukin-2 (IL-2) production, IL-2 receptor alpha-chain (CD25) and beta-chain (CD122) expression, and translocation of PKC beta isozyme in human peripheral blood lymphocytes: evidence for a role of PKC beta 1 in human T cell activation.
Journal of clinical immunology (1994-07-01)
S Aggarwal, S Lee, A Mathur, S Gollapudi, S Gupta
ABSTRACT
To determine a role of protein kinase C (PKC) isozymes in lymphocyte activation, human peripheral blood mononuclear cells were activated with 12-deoxyphorbol-13-O-phenylacetate (dPP; an agonist of both calcium-dependent and calcium-independent PKC isozymes), thymeleatoxin (TX; an activator of calcium-dependent PKC alpha, beta, and gamma), and 12-deoxyphorbol-13-O-phenylacetate 20 acetate (dPPA; an activator of PKC beta 1 isozyme) and examined for DNA synthesis, lymphocyte proliferation, interleukin-2 (IL-2) production, expression of IL-2 receptor alpha and beta chains on CD3+, CD4+, and CD8+ T lymphocytes and CD20+ B lymphocytes, and translocation of PKC beta isozyme from cytosol to membrane fraction. The results show that dPPA activates lymphocytes by inducing the above changes in a manner analogous to that of dPP, TX, and phorbol myristate acetate. These data suggest that PKC beta 1 is involved in the activation of human peripheral blood T and B lymphocytes.
Sigma-Aldrich
dPPA, ≥98% (HPLC)