MilliporeSigma
  • Multiple conformations of the sea anemone polypeptide anthopleurin-A in solution.
Multiple conformations of the sea anemone polypeptide anthopleurin-A in solution.
Protein science : a publication of the Protein Society (1994-07-01)
M J Scanlon, R S Norton
ABSTRACT
Anthopleurin-A (AP-A) is a member of a family of sea anemone-derived polypeptides that interact with sodium channels in a voltage-dependent manner, producing a positive inotropic effect on the mammalian heart. There has been considerable interest in this molecule as a lead compound for the development of novel therapeutic agents. Earlier attempts to define the 3-dimensional structure of AP-A were complicated by the fact that it was found to exist in 2 conformations in solution. Using 1H- and 13C-NMR spectroscopy, we have now shown that this conformational heterogeneity arises from cis-trans isomerization about the Gly 40-Pro 41 peptide bond and that in the major form of the protein this peptide bond adopts a cis conformation. Furthermore, the increased sensitivity afforded by higher-field NMR has allowed identification of additional minor conformations of AP-A, the origin of which is presently unknown. We believe there will be many more examples of the detection by high-field NMR of previously unobserved minor conformations of proteins in solution.