MilliporeSigma
  • [Evaluation of maximal permissible concentration of 2-allyl oxyethanol in the atmospheric air].
[Evaluation of maximal permissible concentration of 2-allyl oxyethanol in the atmospheric air].
Gigiena i sanitariia (1995-03-01)
M A Pinigin, L A Tepikina, A A Safulin, N P Zinovéva, E L Skvortsova, Z V Shipulina, N N Sizova, L F Astakhova
ABSTRACT
The threshold irritable concentration of 2-allyl oxyethanol (2-AOE) in ambient air was determined at the level of 0.68 mg/m3. The threshold of chronic effect of 2-AOE is 0.075 mg/m3. The mean daily permissible concentration of 2-AOE is suggested at the level of 0.01 mg/m3.
Sigma-Aldrich
2-Allyloxyethanol, 98%
Pricing and availability is not currently available.