MilliporeSigma
  • [Diffusional system for examination of the solubility and penetration of water-insoluble compounds (author's transl)].
[Diffusional system for examination of the solubility and penetration of water-insoluble compounds (author's transl)].
Zentralblatt fur Bakteriologie, Parasitenkunde, Infektionskrankheiten und Hygiene. Zweite naturwissenschaftliche Abteilung: Mikrobiologie der Landwirtschaft der Technologie und des Umweltschutzes (1980-01-01)
I L Genz, H D Babenzien, M Köhler
ABSTRACT
A diffusion cell type which is suitable for investigation of water-insoluble compounds is shown. The chamber consists of two compartments, the so-called bottom-chamber, which contains the solid substance under investigation, and the main chamber, which is above it. In this chamber only the dissolved part of the substance is present. The solubility of dibenzylsulfide was determined and it was demonstrated that the dissolved part of this compound is accumulated by a receptor substance from the liquid phase.
Sigma-Aldrich
Dibenzyl sulfide, ≥95.0% (HPLC)