MilliporeSigma
  • Treatment of Trichuris trichiura infection with a single dose of oxantel pamoate.
Treatment of Trichuris trichiura infection with a single dose of oxantel pamoate.
Scandinavian journal of infectious diseases (1982-01-01)
K Peldán, T Pitkänen
ABSTRACT
166 patients with trichuriasis have been treated at the Department of Tropical Diseases, Aurora Hospital, Helsinki, from January 1975 to October 1979. Final results could be confirmed in 141 patients. Thiabendazole was found discouraging, because of frequent side effects and low cure rate (29% of 24 in-patients). In contrast, a single dose of oxantel pamoate gave minimal side effects in 37 in-patients and in 80 outpatients with a high overall cure rate of 98%. Neither allergic nor adverse haematological reactions were encountered during 3 years and 10 months' experience of treatment with oxantel pamoate.
Product Description
Supelco
Oxantel pamoate, analytical standard