MilliporeSigma
  • [Diminution of the rate of growth of HeLa cells caused by thioproline (Tp) and 2-aminothiazoline HCl (2-AT). Effect of L-proline].
[Diminution of the rate of growth of HeLa cells caused by thioproline (Tp) and 2-aminothiazoline HCl (2-AT). Effect of L-proline].
Revista espanola de oncologia (1982-01-01)
J J Díaz Gil, C Trilla
ABSTRACT
Some aspects of the effect of thioproline and 2-amino-thiazoline-HCl on HeLa cell cultures are studied. Both drugs, at 1 mM concentrations, produce morphological changes that become stabilized after four days. The morphology changes depend on the microtubules, microfilaments, and de novo protein synthesis. Both drugs diminish the growth rate of HeLa cells, the effect being partially reverted by high concentrations (2 mM) of L-proline.