MilliporeSigma
  • Formation of novel nitrofuran metabolites in xanthine oxidase-catalyzed reduction of methyl 5-nitro-2-furoate.
Formation of novel nitrofuran metabolites in xanthine oxidase-catalyzed reduction of methyl 5-nitro-2-furoate.
Archives of biochemistry and biophysics (1984-07-01)
H Yamada, K Tatsumi, Y Kawazoe, H Yoshimura
ABSTRACT
After methyl 5-nitro-2-furoate was incubated with milk xanthine oxidase, three reduction products were isolated from the incubation mixture. Among them, two reduction products were new types of nitrofuran metabolites, i.e., metabolites 1 and 2 were identified as the dihydroxyhydrazine derivative (1,2-dihydroxy-1,2-di(5-methoxycarbonyl-2-furyl)hydrazine) and the hydroxylaminofuran derivative (methyl 5-hydroxylamino-2-furoate), respectively. Metabolite 3 was also identified as the aminofuran derivative (methyl 5-amino-2-furoate) by comparison with a synthetic sample.